Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930555B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040633.1 | 4 | 1359 | |||
GenBank mRNA | AK016127.1 | - | 4 | 1359 |