Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crnde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033641.3 | 7 | 1192 | |||
NR_110452.1 | 4 | 1011 | ||||
GenBank mRNA | AK017076.1 | - | 7 | 1192 | ||
CX205694.1 | - | 4 | 722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB751125.1 | - | 3 | 351 | Chr. 8 - 92326038 - 92344943 [-] on Build GRCm38 |