Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1500032P08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005351.1 | - | 1 | 628 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU701317.1 | - | 1 | 36 | Chr. X - 20931519 - 20932166 [-] on Build GRCm38 |