Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm40826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038969.1 | - | 1449 |
Gene Symbol: Sugct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC064756.1 | - | 3 | 1328 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB179576.2 | - | 1 | 623 | Chr. 13 - 17682713 - 17683350 [-] on Build GRCm38 |