Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040588.1 | 4 | 580 | |||
GenBank mRNA | AK019118.1 | - | 4 | 582 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710128.1 | - | 4 | 582 | Chr. 3 - 102720231 - 102735419 [-] on Build GRCm38 |