Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066208.1 | - | 1, 2 | 22 | ||
BC099934.1 | - | 1, 2 | 30 | 6432976 | ||
BC145554.1 | - | 1, 2 | 18 | 9054118 | ||
GQ915612.1 | - | 1, 2 | 45 |