Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A230051N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038632.1 | - | 2 | 446 | ||
AK080548.1 | - | 2 | 487 | |||
BB258200.2 | - | 1 | 407 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB631910.1 | - | 2 | 446 | Chr. 17 - 56241756 - 56242677 [+] on Build GRCm38 |