Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A230051N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038632.1 | - | 2 | 285 | ||
AK080548.1 | - | 2 | 325 | |||
BB258200.2 | - | 1 | 245 |