Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A330049N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040646.1 | 2 | 187 | |||
GenBank mRNA | AK039487.1 | - | 2 | 188 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633452.1 | - | 2 | 188 | Chr. 10 - 72977663 - 73086705 [-] on Build GRCm38 |