Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700065L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108032.1 | 3 | 251 | |||
GenBank mRNA | AK006896.1 | - | 3 | 254 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706888.1 | - | 3 | 252 | Chr. 6 - 73436963 - 73471021 [-] on Build GRCm38 |