Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 1700109G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033788.1 | 3 | 759 | |||
GenBank mRNA | AK007149.1 | - | 3 | 759 | ||
AK018935.1 | - | 3 | 722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU935660.1 | - | 3 | 715 | Chr. 14 - 61292980 - 61305289 [-] on Build GRCm38 |