Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9430018C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020423.1 | - | 1 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721608.1 | - | 1 | 106 | Chr. 9 - 97764508 - 97765155 [-] on Build GRCm38 |