Gene Symbol: Akap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018023.1 | - | 1 | 894 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB668129.1 | - | 1 | 301 | Chr. 7 - 75612437 - 75613059 [+] on Build GRCm38 |