Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700125G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040548.1 | 2 | 459 | |||
GenBank mRNA | AK007279.1 | - | 2 | 459 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707381.1 | - | 466 | Chr. - - [-] on Build |