Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930445N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125720.1 | 2 | 308 | |||
GenBank mRNA | AK015395.1 | - | 2 | 309 |