Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700072G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006937.1 | - | 2 | 276 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706952.1 | - | 2 | 271 | Chr. 1 - 51457932 - 51458931 [-] on Build GRCm38 |