Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank4os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040437.1 | 3 | 896 | |||
GenBank mRNA | AK043321.1 | - | 3 | 897 | ||
AK166708.1 | - | 3 | 867 |