Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm30340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044064.1 | - | 1, 2 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB641981.1 | - | 1, 2 | 110 | Chr. 3 - 17330234 - 17334425 [-] on Build GRCm38 |