Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040741.1 | - | 1 | 861 | ||
AK040882.1 | - | 1 | 889 | |||
AK047674.1 | - | 1 | 878 | |||
AK048101.1 | - | 1 | 1047 | |||
AK084338.1 | - | 1 | 878 | |||
AK164170.1 | - | 1 | 938 | |||
AK169469.1 | - | 1 | 886 | |||
BC063049.1 | - | 1 | 438 | |||
BC080683.1 | - | 1 | 877 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ930208.1 | - | 1 | 60 | Chr. 3 - 89771520 - 89772408 [-] on Build GRCm38 |