Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014549.2 | - | 363 | |||
AK040741.1 | - | 1 | 337 | |||
AK040882.1 | - | 1 | 365 | |||
AK047674.1 | - | 1 | 354 | |||
AK048101.1 | - | 1 | 523 | |||
AK084338.1 | - | 1 | 353 | |||
AK164170.1 | - | 1 | 414 | |||
AK169469.1 | - | 1 | 362 | |||
BC080683.1 | - | 1 | 353 | |||
BC106127.1 | - | 1 | 367 |