Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700120E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045368.1 | 4 | 1907 | |||
GenBank mRNA | AK007214.1 | - | 4 | 1908 |