Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102640923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158984.1 | - | 3 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY285137.1 | - | 3 | 219 | Chr. 8 - 76797053 - 76899985 [-] on Build GRCm38 |