Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007000.1 | - | 464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707056.1 | - | 2 | 464 | Chr. 7 - 19715016 - 19721720 [+] on Build GRCm38 |