Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l4aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038151.1 | 3 | 414 | |||
NR_038152.1 | 2 | 351 | ||||
GenBank mRNA | AA269689.1 | - | 3 | 355 | ||
AK010403.1 | - | 3 | 393 | |||
BY266174.1 | - | 3 | 413 |