Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l4aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038151.1 | 1 | 69 | |||
NR_038152.1 | 1 | 69 | ||||
GenBank mRNA | AA269689.1 | - | 1 | 13 | ||
AK010403.1 | - | 1 | 48 | |||
BY266174.1 | - | 1 | 69 |