Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 4930577N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073429.1 | 1 | 1440 | |||
GenBank mRNA | AK016297.1 | - | 1 | 1440 | ||
AK017883.1 | - | 1 | 1309 | |||
AK086057.1 | - | 1 | 1385 |