Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610019N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011471.1 | - | 1 | 143 | ||
AK013186.1 | - | 1 | 99 |
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY452064.1 | - | 77 |