Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137400.1 | - | 1 | 2789 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB130570.2 | - | 1 | 194 | Chr. 15 - 102590321 - 102591009 [-] on Build GRCm38 |