Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040496.1 | 2 | 141 | |||
GenBank mRNA | AK005124.1 | - | 2 | 142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705319.1 | - | 2 | 139 | Chr. 2 - 70489958 - 70563355 [-] on Build GRCm38 |