Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5930409G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020028.1 | - | 1 | 61 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718015.1 | - | 1 | 61 | Chr. 3 - 45274066 - 45274967 [-] on Build GRCm38 |