Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933430M04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045857.2 | 4 | 364 | |||
GenBank mRNA | AK016999.1 | - | 4 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716684.1 | - | 4 | 365 | Chr. 11 - 24481909 - 24569952 [+] on Build GRCm38 |