Gene Symbol: A530065N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046142.1 | 3 | 566 | |||
GenBank mRNA | AK080130.1 | - | 3 | 567 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB637326.1 | - | 3 | 567 | Chr. 13 - 60029711 - 60113578 [+] on Build GRCm38 |