Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210039B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044985.1 | 4 | 584 | |||
GenBank mRNA | AK008780.1 | - | 4 | 590 |
Gene Symbol: A630031M23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041714.1 | - | 1 | 1065 |
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110345.1 | 11 | 2872 | |||
GenBank mRNA | BC059822.1 | - | 11 | 2825 | 6844585 |
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052313.1 | - | 1 | 1973 |
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133564.1 | - | 4 | 2638 | ||
CA319782.1 | - | 1 | 717 |