Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930455D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045381.1 | 2 | 1025 | |||
GenBank mRNA | AK015461.1 | - | 2 | 1026 |