Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHLAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104319.1 | 3 | 607 | |||
NR_104320.1 | 3 | 607 | ||||
NR_104322.1 | 5 | 846 | ||||
NR_104323.1 | 4 | 745 | ||||
NR_104324.1 | 4 | 708 | ||||
NR_104325.1 | 4 | 708 | ||||
GenBank mRNA | JX117418.1 | - | 3 | 607 | ||
JX117419.1 | - | 3 | 607 | |||
JX117421.1 | - | 5 | 846 | |||
JX117422.1 | - | 4 | 745 | |||
JX117423.1 | - | 4 | 708 | |||
JX117424.1 | - | 4 | 708 |