Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024124.2 | 2 | 597 | |||
NR_024125.2 | 3 | 873 | ||||
NR_024126.1 | 3 | 636 | ||||
GenBank mRNA | AK309872.1 | - | 3 | 871 | ||
BC007373.2 | - | 3 | 636 | |||
BC018892.2 | - | 3 | 636 | |||
BC098428.1 | - | 3 | 873 |