Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC151121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122040.1 | 5 | 1422 | |||
NR_122041.1 | 4 | 1334 | ||||
NR_122042.1 | 4 | 1345 | ||||
GenBank mRNA | AK056598.1 | - | 5 | 1379 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000375987.3 | - | 1379 | Chr. 2 - 129242388 - 129242693 [-] on Build GRCh38 |