Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC100507461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134937.1 | 2 | 181 | |||
GenBank mRNA | AA393442.1 | - | 2 | 183 | ||
BX096822.1 | - | 2 | 136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000490465.1 | - | 2 | 181 | Chr. 3 - 148078159 - 148088029 [+] on Build GRCh38 |