Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120458.1 | 4 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000536112.1 | - | 2 | 229 | Chr. 12 - 68431855 - 68432654 [-] on Build GRCh38 |