Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 3 | 3043 | |||
GenBank mRNA | AK123643.1 | - | 3 | 3043 | ||
BC132803.1 | - | 3 | 783 | |||
BC132805.1 | - | 3 | 783 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000373171.2 | - | 1 | 474 | Chr. 6 - 40344345 - 40356006 [-] on Build GRCh38 |