Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC93463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135202.1 | 5 | 991 | |||
GenBank mRNA | AL122100.1 | - | 5 | 991 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000244820.2 | - | 991 | Chr. 2 - 237055427 - 237056166 [-] on Build GRCh38 |