Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 513 | |||
NR_028508.1 | 1 | 294 | ||||
NR_028509.1 | 1 | 294 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 294 | ||
AL050198.1 | - | 1 | 37 | |||
BC069304.1 | - | 1 | 294 | |||
BC096696.1 | - | 1 | 294 | |||
BC096727.1 | - | 1 | 294 | |||
BC098111.1 | - | 1 | 294 |