Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 6 | 705 | |||
GenBank mRNA | AF277181.1 | - | 6 | 493 | ||
AK092096.1 | - | 3 | 1874 | |||
BC022205.1 | - | 6 | 1334 | |||
BC034611.1 | - | 4 | 1556 | |||
BC045792.1 | - | 5 | 1064 | |||
BC064589.1 | - | 4 | 1595 | |||
BC068048.1 | - | 6 | 705 | |||
BC107730.1 | - | 3 | 1097 | |||
KJ782215.1 | - | 7 | 1834 |