Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173600.2 | NP_775871.2 | 21982 | |||
GenBank mRNA | CR749517.1 | - | 10 | 3090 | ||
HM801842.1 | - | 21982 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000474954.5 | - | 10 | 3089 | Chr. 12 - 40522485 - 40534803 [+] on Build GRCh38 |