Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105377213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135614.1 | 2 | 159 | |||
GenBank mRNA | BX103087.1 | - | 2 | 159 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454307.1 | - | 2 | 159 | Chr. X - 151515551 - 151516245 [+] on Build GRCh38 |