Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123065.1 | NP_001116537.1 | 3 | 2707 | ||
GenBank mRNA | AK126096.1 | - | 3 | 2715 |