Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110204.1 | 3 | 783 | |||
NR_110205.1 | 3 | 627 | ||||
NR_110206.1 | 2 | 562 | ||||
GenBank mRNA | AL520497.3 | - | 3 | 448 | ||
BI915242.1 | - | 2 | 224 | |||
BX538254.1 | - | 3 | 874 | |||
DT216517.1 | - | 3 | 336 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415236.1 | - | 3 | 461 | Chr. 2 - 178414023 - 178432282 [+] on Build GRCh38 |