Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033979.2 | 3 | 718 | |||
NR_110458.1 | 3 | 992 | ||||
NR_110459.1 | 5 | 943 | ||||
NR_110460.1 | 5 | 913 | ||||
NR_110461.1 | 4 | 848 | ||||
NR_110462.1 | 2 | 829 | ||||
NR_110463.1 | 4 | 811 | ||||
NR_110464.1 | 4 | 799 | ||||
NR_110465.1 | 4 | 797 | ||||
NR_110466.1 | 3 | 727 | ||||
NR_110467.1 | 3 | 700 | ||||
NR_110468.1 | 3 | 688 | ||||
GenBank mRNA | BC030713.2 | - | 3 | 979 | ||
BC047481.1 | - | 3 | 979 |