Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033979.2 | 3 | 436 | |||
NR_110458.1 | 3 | 710 | ||||
NR_110459.1 | 5 | 661 | ||||
NR_110460.1 | 5 | 631 | ||||
NR_110461.1 | 4 | 566 | ||||
NR_110462.1 | 2 | 547 | ||||
NR_110463.1 | 4 | 529 | ||||
NR_110464.1 | 4 | 517 | ||||
NR_110465.1 | 4 | 515 | ||||
NR_110466.1 | 3 | 445 | ||||
NR_110467.1 | 3 | 418 | ||||
NR_110468.1 | 3 | 406 | ||||
GenBank mRNA | BC030713.2 | - | 3 | 697 | ||
BC047481.1 | - | 3 | 697 | |||
CB153479.1 | - | 3 | 436 | |||
DA326484.1 | - | 5 | 621 | |||
DA398428.1 | - | 4 | 422 | |||
DA736032.1 | - | 4 | 567 | |||
DB045673.1 | - | 3 | 373 | |||
DB067012.1 | - | 3 | 402 | |||
DB100342.1 | - | 4 | 483 | |||
DB100521.1 | - | 4 | 430 | |||
HY138745.1 | - | 3 | 447 |