Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015429.1 | 8 | 960 | |||
NR_024429.1 | 8 | 1056 | ||||
NR_024431.2 | 10 | 1252 | ||||
GenBank mRNA | AK097923.1 | - | 4 | 1671 | ||
AK098249.1 | - | 10 | 1697 | |||
BC102001.1 | - | 7 | 803 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456353.5 | - | 10 | 1699 | Chr. 10 - 78943328 - 79067895 [-] on Build GRCh38 |